Merck
  • Enhanced urothelial ATP release and contraction following intravesical treatment with the cytotoxic drug, doxorubicin.
Enhanced urothelial ATP release and contraction following intravesical treatment with the cytotoxic drug, doxorubicin.
Naunyn-Schmiedeberg's archives of pharmacology (2015-02-17)
Sung-Hyun Kang, Catherine McDermott, Stefanie Farr, Russ Chess-Williams
ABSTRACT
Intravesical administration of the cytotoxic drug doxorubicin is a common treatment for superficial carcinoma of the bladder, but it is associated with significant urological adverse effects. The aim of this study was to identify doxorubicin-induced changes in the local mechanisms involved in regulating bladder function. As a model of intravesical doxorubicin administration in patients, doxorubicin (1 mg/mL) was applied to the luminal surface of porcine bladders for 60 min. Following treatment, the release of urothelial/lamina propria mediators (acetylcholine (Ach), ATP and prostaglandin E2 (PGE2) and contractile responses of isolated tissue strips was investigated. Doxorubicin pretreatment did not affect contractile responses of detrusor muscle to carbachol, but did enhance neurogenic detrusor responses to electrical field stimulation (219 % at 5 Hz). Contractions of isolated strips of urothelium/lamina propria to carbachol were also enhanced (30 %) in tissues from doxorubicin pretreated bladders. Isolated strips of urothelium/lamina propria from control bladders demonstrated a basal release of all three mediators (Ach > ATP > PGE2), with increased release of ATP when tissues were stretched. In tissues from doxorubicin-pretreated bladders, the basal release of ATP was significantly enhanced (sevenfold), while the release of acetylcholine and PGE2 was not affected. The application of luminal doxorubicin, under conditions that mimic intravesical administration to patients, affects urothelial/lamina propria function (increased contractile activity and ATP release) and enhances efferent neurotransmission without affecting detrusor smooth muscle. These actions would enhance bladder contractile activity and sensory nerve activity and may explain the adverse urological effects observed in patients following intravesical doxorubicin treatment.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis