Merck
  • Feasibility of a facile butanol bioproduction using planetary mill pretreatment.
Feasibility of a facile butanol bioproduction using planetary mill pretreatment.
Bioresource technology (2015-09-16)
Jeong Heo Kwon, Hyunsoo Kang, Byoung-In Sang, Yunje Kim, Jiho Min, Robert J Mitchell, Jin Hyung Lee
ABSTRACT
A facile butanol bioproduction process was developed using planetary milling, and Pinus rigida wood waste as a model substrate for fermentable sugars. The use of planetary milling as the pretreatment eliminates the need for washing and transfer of the biomass prior to enzymatic hydrolysis. Moreover, using this pretreatment process resulted in the production of only 0.072 ± 0.003 g/L soluble phenolic compounds, a concentration that was not inhibitory towards Clostridium beijerinckii NCIMB 8052. As the milling was performed in a compatible buffer (50mM acetate, pH 4.8), the enzymatic hydrolysis step was initiated by simply adding the cellulase cocktail powder directly to pretreated biomass without washing the biomass or exchanging the buffer, resulting in a glucose yield of 31 g/L (84.02%). Fermentation of the hydrolysate samples by C. beijerinckii NCIMB 8052 gave slightly better butanol yields than cultures grown in a typical lab media (P2), with final concentrations of 6.91 and 6.66 g/L, respectively.
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)