Merck
  • Metabolic faecal fingerprinting of trans-resveratrol and quercetin following a high-fat sucrose dietary model using liquid chromatography coupled to high-resolution mass spectrometry.
Metabolic faecal fingerprinting of trans-resveratrol and quercetin following a high-fat sucrose dietary model using liquid chromatography coupled to high-resolution mass spectrometry.
Food & function (2015-07-15)
Usune Etxeberria, Noemi Arias, Noemí Boqué, Ana Romo-Hualde, M Teresa Macarulla, María P Portillo, Fermín I Milagro, J Alfredo Martínez
ABSTRACT
Faecal non-targeted metabolomics deciphers metabolic end-products resulting from the interactions among food, host genetics, and gut microbiota. Faeces from Wistar rats fed a high-fat sucrose (HFS) diet supplemented with trans-resveratrol and quercetin (separately or combined) were analysed by liquid chromatography coupled to high-resolution mass spectrometry (LC-HRMS). Metabolomics in faeces are categorised into four clusters based on the type of treatment. Tentative identification of significantly differing metabolites highlighted the presence of carbohydrate derivatives or conjugates (3-phenylpropyl glucosinolate and dTDP-D-mycaminose) in the quercetin group. The trans-resveratrol group was differentiated by compounds related to nucleotides (uridine monophosphate and 2,4-dioxotetrahydropyrimidine D-ribonucleotide). Marked associations between bacterial species (Clostridium genus) and the amount of some metabolites were identified. Moreover, trans-resveratrol and resveratrol-derived microbial metabolites (dihydroresveratrol and lunularin) were also identified. Accordingly, this study confirms the usefulness of omics-based techniques to discriminate individuals depending on the physiological effect of food constituents and represents an interesting tool to assess the impact of future personalized therapies.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid