Merck
  • Functional characteristics of L1156F-CFTR associated with alcoholic chronic pancreatitis in Japanese.
Functional characteristics of L1156F-CFTR associated with alcoholic chronic pancreatitis in Japanese.
American journal of physiology. Gastrointestinal and liver physiology (2015-06-20)
Shiho Kondo, Kotoyo Fujiki, Shigeru B H Ko, Akiko Yamamoto, Miyuki Nakakuki, Yasutomo Ito, Nikolay Shcheynikov, Motoji Kitagawa, Satoru Naruse, Hiroshi Ishiguro
ABSTRACT
Although cystic fibrosis is rare in Japanese, measurement of sweat Cl(-) has suggested mild dysfunction of cystic fibrosis transmembrane conductance regulator (CFTR) in some patients with chronic pancreatitis. In the present study, we have investigated the association of CFTR variants and chronic pancreatitis in Japanese and the functional characteristics of a Japanese- and pancreatitis-specific CFTR variant, L1156F. Seventy patients with alcoholic chronic pancreatitis, 18 patients with idiopathic chronic pancreatitis, and 180 normal subjects participated. All exons and their boundaries and promoter region of the CFTR gene were sequenced. Human embryonic kidney-293 cells were transfected with three CFTR variants (M470V, L1156F, and M470V+L1156F), and the protein expression was examined. Xenopus laevis oocytes were injected with the CFTR variants, and bicarbonate (HCO3 (-)) transport activity was examined. CFPAC-1 cells were transfected with the CFTR variants and Cl(-)/HCO3 (-) exchange activity was examined. Six variants (E217G, I556V, M470V, L1156F, Q1352H, and R1453W) were identified in the coding region of the CFTR gene. Cystic fibrosis-causing mutations were not found. The allele frequencies of L1156F and Q1352H in alcoholic chronic pancreatitis (5.0 and 7.9%) were significantly (P < 0.01) higher than those in normal subjects (0.6 and 1.9%). L1156F was linked with a worldwide CFTR variant, M470V. Combination of M470V and L1156F significantly reduced CFTR expression to ∼60%, impaired CFTR-mediated HCO3 (-)/Cl(-) transport activity to 50-60%, and impaired CFTR-coupled Cl(-)/HCO3 (-) exchange activity to 20-30%. The data suggest that the Japanese-specific CFTR variant L1156F causes mild dysfunction of CFTR and increases the risk of alcoholic chronic pancreatitis in Japanese.
Sigma-Aldrich
Tridodecylamine, ≥97.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O