Merck
  • Characterization of five passive sampling devices for monitoring of pesticides in water.
Characterization of five passive sampling devices for monitoring of pesticides in water.
Journal of chromatography. A (2015-06-20)
Lutz Ahrens, Atlasi Daneshvar, Anna E Lau, Jenny Kreuger
ABSTRACT
Five different passive sampler devices were characterized under laboratory conditions for measurement of 124 legacy and current used pesticides in water. In addition, passive sampler derived time-weighted average (TWA) concentrations were compared to time-integrated active sampling in the field. Sampling rates (RS) and passive sampler-water partition coefficients (KPW) were calculated for individual pesticides using silicone rubber (SR), polar organic chemical integrative sampler (POCIS)-A, POCIS-B, Chemcatcher(®) SDB-RPS and Chemcatcher(®) C18. The median RS (Lday(-1)) decreased as follows: SR (0.86)>POCIS-B (0.22)>POCIS-A (0.18)>Chemcatcher(®) SDB-RPS (0.05)>Chemcatcher(®) C18 (0.02), while the median logKPW (Lkg(-1)) decreased as follows: POCIS-B (4.78)>POCIS-A (4.56)>Chemcatcher(®) SDB-RPS (3.17)>SR (3.14)>Chemcatcher(®)C18 (2.71). The uptake of the selected compounds depended on their physicochemical properties, i.e. SR showed a better uptake for more hydrophobic compounds (log octanol-water partition coefficient (KOW)>5.3), whereas POCIS-A, POCIS-B and Chemcatcher(®) SDB-RPS were more suitable for hydrophilic compounds (logKOW<0.70). Overall, the comparison between passive sampler and time-integrated active sampler concentrations showed a good agreement and the tested passive samplers were suitable for capturing compounds with a wide range of KOW's in water.
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis