Merck
  • (211)Astatine-Conjugated Monoclonal CD45 Antibody-Based Nonmyeloablative Conditioning for Stem Cell Gene Therapy.
(211)Astatine-Conjugated Monoclonal CD45 Antibody-Based Nonmyeloablative Conditioning for Stem Cell Gene Therapy.
Human gene therapy (2015-04-29)
Christopher R Burtner, Devikha Chandrasekaran, Erlinda B Santos, Brian C Beard, Jennifer E Adair, Donald K Hamlin, D Scott Wilbur, Brenda M Sandmaier, Hans-Peter Kiem
ABSTRACT
Most hematopoietic stem cell gene therapy studies require host conditioning to allow for efficient engraftment of gene-modified cells. Conditioning regimens with lower treatment-related toxicities are especially relevant for the treatment of nonmalignant blood disorders, such as hemoglobinopathies and immunodeficiencies, and for patients who are otherwise ineligible for conventional high-dose conditioning. Radioimmunotherapy, which employs an α- or a β-emitting radionuclide conjugated to a targeting antibody, is effective for delivering cytotoxic doses of radiation to a cell type of interest while minimizing off-target toxicity. Here, we demonstrate the feasibility of using a nonmyeloablative dose of a monoclonal anti-CD45 antibody conjugated to the α-emitter Astatine-211 ((211)At) to promote engraftment of an autologous gene-modified stem cell graft in the canine model. The doses used provided myelosuppression with rapid autologous recovery and minimal off-target toxicity. Engraftment levels were low in all dogs and reflected the low numbers of gene-modified cells infused. Our data suggest that a cell dose exceeding 1×10(6) cells/kg be used with nonmyeloablative doses of (211)At-anti-CD45 monoclonal antibodies for sustained engraftment in the dog model.
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
TPO human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)