Merck
  • Bond-based bilinear indices for computational discovery of novel trypanosomicidal drug-like compounds through virtual screening.
Bond-based bilinear indices for computational discovery of novel trypanosomicidal drug-like compounds through virtual screening.
European journal of medicinal chemistry (2015-04-18)
Juan Alberto Castillo-Garit, Oremia del Toro-Cortés, Maria C Vega, Miriam Rolón, Antonieta Rojas de Arias, Gerardo M Casañola-Martin, José A Escario, Alicia Gómez-Barrio, Yovani Marrero-Ponce, Francisco Torrens, Concepción Abad
ABSTRACT
Two-dimensional bond-based bilinear indices and linear discriminant analysis are used in this report to perform a quantitative structure-activity relationship study to identify new trypanosomicidal compounds. A data set of 440 organic chemicals, 143 with antitrypanosomal activity and 297 having other clinical uses, is used to develop the theoretical models. Two discriminant models, computed using bond-based bilinear indices, are developed and both show accuracies higher than 86% for training and test sets. The stochastic model correctly indentifies nine out of eleven compounds of a set of organic chemicals obtained from our synthetic collaborators. The in vitro antitrypanosomal activity of this set against epimastigote forms of Trypanosoma cruzi is assayed. Both models show a good agreement between theoretical predictions and experimental results. Three compounds showed IC50 values for epimastigote elimination (AE) lower than 50 μM, while for the benznidazole the IC50 = 54.7 μM which was used as reference compound. The value of IC50 for cytotoxicity of these compounds is at least 5 times greater than their value of IC50 for AE. Finally, we can say that, the present algorithm constitutes a step forward in the search for efficient ways of discovering new antitrypanosomal compounds.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Resazurin sodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
2-Chlorophenol, 98%
Sigma-Aldrich
Chlorophenol Red-β-D-galactopyranoside, ≥90% (HPLC)
Sigma-Aldrich
2-Chlorophenol, ≥99%