Merck
  • Activated and inactivated immune responses in Caenorhabditis elegans against Photorhabdus luminescens TT01.
Activated and inactivated immune responses in Caenorhabditis elegans against Photorhabdus luminescens TT01.
SpringerPlus (2014-10-04)
Kazuki Sato, Toyoshi Yoshiga, Koichi Hasegawa
ABSTRACT
The Gram-negative bacterium Photorhabdus luminescens which symbiotically associates with the entomopathogenic nematode Heterorhabditis bacteriophora, has a broad insecticidal and nematicidal activity. The virulence of P. luminescens toward the non-mutualistic nematode Caenorhabditis elegans has not been described. We showed that when fed on P. luminescens, the intestinal cells of C. elegans worms become delicate and some crystal-like structure was developed within the intestinal lumen. Next, we examined the requirement of the p38 mitogen-activated protein kinase (MAPK) and insulin/IGF-1 signaling pathway against P. luminescens. Depletion of pmk-1 by RNAi enhances susceptibility to P. luminescens, and numerous downstream targets regulated by the p38 MAPK pathway were induced when fed on P. luminescens. On the other hand, knockdown of daf-16 has no effects on C. elegans lifespan, but knockdown of daf-2 dramatically increased resistance to P. luminescens in a daf-16-dependent manner. We also revealed one of the daf-2 ligands ins-7 was induced and ins-7 deletion mutant survived longer when fed on P. luminescens. These results suggest the p38 MAPK pathway is activated and required for the host defense against P. luminescens. Insulin/IGF-1 signaling pathway is inactivated by P. luminescens through the overexpression of insulin-like gene.