Merck
  • Downregulating HMGA2 attenuates epithelial-mesenchymal transition-induced invasion and migration in nasopharyngeal cancer cells.
Downregulating HMGA2 attenuates epithelial-mesenchymal transition-induced invasion and migration in nasopharyngeal cancer cells.
Biochemical and biophysical research communications (2015-05-31)
You-You Xia, Li Yin, Ning Jiang, Wen-Jie Guo, Hao Tian, Xue-Song Jiang, Jing Wu, Meng Chen, Jian-Zhong Wu, Xia He
ABSTRACT
Epithelial-mesenchymal transition (EMT) is associated with invasion and metastasis of cancer cells. High-mobility group AT-hook 2 (HMGA2) has been found to play a critical role in EMT in a number of malignant tumors. However, whether HMGA2 regulates the EMT in human nasopharyngeal carcinoma (NPC) is unclear. The aim of this study was to investigate the effect and mechanism of HMGA2 in inducing invasion and migration in NPC. In NPC tissues samples, the association of HMGA2 mRNA expression with clinicopathological characteristics were estimated by real-time quantitative RT-PCR(qRT-PCR). In vitro, following the silencing of HMGA2 in CNE-1 and CNE-2 cell lines, the viability and metastatic ability were analyzed using Cell Counting Kit-8 (CCK8), colony formation assay, and transwell assay. EMT and transforming growth factor-beta (TGFβ)/Smad3 signaling pathway-related protein expression changes were evaluated using western blot. HMGA2 was upregulated in NPC cell lines and clinical specimens (P < 0.01), and HMGA2 expression correlated significantly with metastasis (P = 0.02) and disease-free survival of NPC (hazard ratio: 3.52; 95% confidence interval: 1.34-7.79; P = 0.01). In addition, following in vitro knockdown of HMGA2, the aggressiveness of cells was markedly inhibited, Vimentin and Snail level was downregulated and E-cadherin expression was upregulated. Moreover, the expression of key proteins TGFβRII and p-Smad3 of the TGFβ/Smad3 signaling pathway was inhibited by the downregulation of HMGA2. HMGA2 might maintain EMT-induced invasion and migration through the TGFβ/Smad3 signaling pathway in NPC cell lines.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hmga2
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)