Merck
  • Hierarchically Structured Hole Transport Layers of Spiro-OMeTAD and Multiwalled Carbon Nanotubes for Perovskite Solar Cells.
Hierarchically Structured Hole Transport Layers of Spiro-OMeTAD and Multiwalled Carbon Nanotubes for Perovskite Solar Cells.
ChemSusChem (2015-05-28)
Jiyong Lee, Mini Mol Menamparambath, Jae-Yeol Hwang, Seunghyun Baik
ABSTRACT
The low electrical conductivity of spiro-OMeTAD hole transport layers impedes further enhancements of the power conversion efficiency (PCE) of perovskite solar cells. We embedded multiwalled carbon nanotubes (MWNTs) in spiro-OMeTAD (spiro-OMeTAD/MWNTs) to increase carrier mobility and conductivity. However, direct electrical contact between CH3 NH3 PbI3 and the MWNTs created pathways for undesirable back-electron transfer, owing to the large work function of MWNTs, limiting enhancements of the PCE. A hierarchical structure of pure spiro-OMeTAD and spiro-OMeTAD/MWNTs was designed to block back-electron transfer and fully exploit the enhanced charge transport of spiro-OMeTAD/MWNTs. The enhanced fill factor, short-circuit current density, open-circuit voltage, and PCE (15.1 %) were achieved by using this hierarchical hole transport layer structure (MWNT concentration=2 wt %). The perovskite solar cells were fabricated by a low-temperature solution process, further decreasing their per-Watt cost.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
4-tert-Butylpyridine, 98%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile