Merck
  • YIP1 family member 4 (YIPF4) is a novel cellular binding partner of the papillomavirus E5 proteins.
YIP1 family member 4 (YIPF4) is a novel cellular binding partner of the papillomavirus E5 proteins.
Scientific reports (2015-08-04)
Marietta Müller, Christopher W Wasson, Ramya Bhatia, Sally Boxall, David Millan, Grace Y S Goh, Jürgen Haas, Nicola J Stonehouse, Andrew Macdonald
ABSTRACT
E5 proteins are amongst the least understood of the Human Papillomavirus (HPV) encoded gene products. They are small, membrane-integrated proteins known to modulate a number of critical host pathways associated with pathogenesis including growth factor receptor signaling and immune evasion. Their role in the virus life cycle is less clear, indicating a role in the productive stages of the life cycle. However, a mechanism for this is currently lacking. Here we describe the identification of a novel binding partner of E5, YIPF4 using yeast two-hybrid analysis. YIPF4 is also a poorly characterized membrane spanning protein. Mutagenesis studies implicated the transmembrane regions of each protein as important for their interaction. Binding to YIPF4 was found for all E5 proteins tested suggesting that this interaction may mediate a conserved E5 function. In normal human keratinocytes YIPF4 expression was down-regulated upon differentiation and this reduction was partially rescued in cells harbouring HPV. Despite the conserved nature of the interaction with E5, siRNA mediated depletion of YIPF4 failed to impede two well-characterized functions of E5, namely EGFR trafficking or HLA class I presentation. Continued studies of YIPF4 are warranted to determine its role in the PV life cycle.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Pricing and availability is not currently available.