Merck
  • Sodium polyanethol sulfonate (SPS) falsifies protein staining and quantification and how to solve this problem.
Sodium polyanethol sulfonate (SPS) falsifies protein staining and quantification and how to solve this problem.
Journal of microbiological methods (2015-10-13)
Marcel Prax, Shideh Vatani Shahmirzadi, Friedrich Götz
ABSTRACT
Sodium polyanethol sulfonate (SPS) is an anionic detergent with a broad range of activities and applications. While studying the excretion of cytoplasmic proteins in Staphylococcus aureus SPS was used as cell lysis inhibitor. When investigating the protein pattern of culture supernatants from cells grown in the absence or presence of SPS by Coomassie blue stained polyacrylamide gel the amount of protein bands was significantly decreased in the presence of SPS, suggesting that this effect was due to inhibition of cell lysis. However, various control studies showed that the apparent decreased protein secretion was an artifact due to the interference of SPS with Coomassie blue- and silver-staining. The only alternative method that was uninfluenced by SPS was imidazole-SDS-zinc staining. This is the method of choice particularly when protein interfering compounds are present in the extracts. For protein quantification in liquid samples the bicinchoninic acid (BCA) assay appeared to be the method of choice in the presence of SPS. The assay is based on neutral peptide bonds and is therefore rather insensitive to interfering compounds. This study shows that SPS and most likely also related detergents might falsify conventional protein staining and quantification methods.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)