Merck
  • T helper 9 cells induced by plasmacytoid dendritic cells regulate interleukin-17 in multiple sclerosis.
T helper 9 cells induced by plasmacytoid dendritic cells regulate interleukin-17 in multiple sclerosis.
Clinical science (London, England : 1979) (2015-02-24)
Gabriella Ruocco, Silvia Rossi, Caterina Motta, Giulia Macchiarulo, Francesca Barbieri, Marco De Bardi, Giovanna Borsellino, Annamaria Finardi, Maria Grazia Grasso, Serena Ruggieri, Claudio Gasperini, Roberto Furlan, Diego Centonze, Luca Battistini, Elisabetta Volpe
ABSTRACT
Multiple sclerosis (MS) is a chronic disease of the central nervous system (CNS) characterized by persistent inflammation orchestrated by cluster of differentiation (CD) 4 T helper (Th) cells. In particular, Th1 and Th17 cells amplify, whereas T regulatory (Treg) cells moderate inflammation. The role of other Th subsets in MS is not clear. In the present study, we investigated the generation of different Th responses by human dendritic cells (DCs) in MS. We compared the production of several Th cytokines by naive CD4+ T-cells polarized with myeloid and plasmacytoid DCs (mDCs and pDCs) in healthy donors (HD) and relapsing-remitting (RR)-MS patients. We found that resiquimod-stimulated mDCs were able to activate Th17 differentiation, whereas pDCs induced interleukin (IL)-10-producing Th cells. Surprisingly, resiquimod-stimulated pDCs from MS patients also significantly induced the differentiation of Th9 cells, which produce IL-9 and are known to be involved in allergic diseases. We investigated the potential role of IL-9 in MS. We found that IL-9 activated signal transducer and activator of transcription (STAT) 1 and STAT5 phosphorylation and interfered with IL-17 and interferon (IFN) regulatory transcription factor (IRF)-4 expression in Th17-polarized cells. Moreover, in the cerebrospinal fluid (CSF) of 107 RR-MS patients, IL-9 inversely correlated with indexes of inflammatory activity, neurodegeneration and disability progression of MS. High levels of IL-9 were associated with the absence of IL-17 in the CSF of RR-MS patients. Our results demonstrate a Th9-inducing potential of pDCs in MS, suggesting an immunoregulatory role leading to attenuation of the exaggerated Th17 inflammatory response.
Sigma-Aldrich
IL-9 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Pricing and availability is not currently available.
Sigma-Aldrich
IL-9 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Pricing and availability is not currently available.