Merck
  • Effects of Long-Term Antiglaucoma Eye Drops on Conjunctival Structures: An In Vivo Confocal Microscopy Study.
Effects of Long-Term Antiglaucoma Eye Drops on Conjunctival Structures: An In Vivo Confocal Microscopy Study.
Journal of ophthalmology (2015-07-15)
Wenqing Zhu, Xiangmei Kong, Jianjiang Xu, Xinghuai Sun
ABSTRACT
Purpose. The study was aimed at comparing the long-term effects of different antiglaucoma eye drops on conjunctival structures using laser scanning confocal microscopy. Methods. Eighty patients diagnosed with primary open-angle glaucoma and twenty healthy volunteers were included in this study. The participants were divided into 5 groups according to the different medications. The lachrymal film break-up time, Schirmer's I test, and Ocular Surface Disease Index Questionnaire were performed in all subjects. The confocal microscopy was used to observe the basal epithelial cell density (ECD), goblet cell density (GCD), dendritic cell density (DCD), and subepithelial collagen fiber diameter (SFD). Results. Statistically significant differences were found among the control group and the antiglaucoma therapy groups in the values of three clinical data (P < 0.05). The GCD, DCD, and SFD showed significant differences in all glaucoma groups when compared to the control (P < 0.001). Moreover, the prostaglandin group differed from the other antiglaucoma therapy groups in the GCD and SFD (P < 0.05). Conclusions. Our study confirmed the significant differences in the conjunctival structures based on the effects of antiglaucoma medications. Less pronounced changes were found in the patients treated with prostaglandin analogue than in the other kinds of antiglaucoma therapies.
Sigma-Aldrich
UK 14,304
Sigma-Aldrich
Fluprostenol isopropyl ester, ≥98%, ethanol solution