Merck
Antitcoagulant and antiplatelet activities of scolymoside.
BMB reports (2015-04-19)
Eun-Kyung Yoon, Sae-Kwang Ku, Wonhwa Lee, Soyoung Kwak, Hyejin Kang, Byeongjin Jung, Jong-Sup Bae
ABSTRACT
Cyclopia subternata is a medicinal plant commonly used in traditional medicine to relieve pain. Here, the anticoagulant effects of scolymoside, an active compound in C. subternata, were examined by monitoring activated partial thromboplastin time (aPTT), prothrombin time (PT), and the activities of thrombin and activated factor X (FXa). The effects of scolymoside on plasminogen activator inhibitor type 1 (PAI-1) and tissue-type plasminogen activator (t-PA) expression were evaluated in tumor necrosis factor (TNF)-α-activated human endothelial cells. Treatment with scolymoside resulted in prolonged aPTT and PT and the inhibition of thrombin and FXa activities and production. In addition, scolymoside inhibited thrombin-catalyzed fibrin polymerization and platelet aggregation. Scolymoside also elicited anticoagulant effects in mice, including a significant reduction in the PAI-1 to t-PA ratio. Collectively, these findings indicate that scolymoside possesses anticoagulant activities and could be developed as a novel anticoagulant.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Luteolin 7-rutinoside, ≥95% (LC/MS-ELSD)