Merck
  • Crystal Structure of Mg(2+) Containing Hemopexin-Fold Protein from Kabuli Chana (Chickpea-White, CW-25) at 2.45 Å Resolution Reveals Its Metal Ion Transport Property.
Crystal Structure of Mg(2+) Containing Hemopexin-Fold Protein from Kabuli Chana (Chickpea-White, CW-25) at 2.45 Å Resolution Reveals Its Metal Ion Transport Property.
The protein journal (2015-08-06)
Suresh Kumar, Avinash Singh, Shavait Yamini, Sharmistha Dey, T P Singh
ABSTRACT
Plant seeds contain a number of proteins which play important roles in the protection and the process of germination of seeds. We have isolated and purified a 25 kDa protein from Kabuli Chana (Cicer arietinum L., Chickpea-white, CW-25). The CW-25 protein was crystallized using 0.5 M magnesium acetate, 0.1 M sodium cacodylate and 20 % (w/v) polyethylene glycol 8000, pH 6.5. The crystals of CW-25 belonged to space group P3 with unit cell dimensions, a = b = 80.5 Å, and c = 69.2 Å. The structure of CW-25 was determined using molecular replacement method and refined to an R factor of 0.152. The buried surface area between two molecules was found to be approximately 653 Å(2) indicating the formation of a weak homodimer. The polypeptide chain of CW-25 adopted a hemopexin-fold with four-bladed β-propellers. The structure formed a central tunnel-like architecture. A magnesium ion was observed in the centre of the tunnel. It was located at distances varying between 2.3 and 2.7 Å from five oxygen atoms of which four were backbone oxygen atoms belonging to residues, Asn7, Asp65, Asp121 and Asp174 while the fifth oxygen atom, O(δ1) was from the side chain of Asn7. The approximate length of the tunnel was 30 Å. Furthermore, a series of carbonyl oxygen atoms were present along the internal face of the tunnel. The diameter of the tunnel varied from 4.6 to 6.2 Å. The diameter and chemical environment of the tunnel clearly indicated that it might be used for the transport of various metal ions across the molecule.
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)