Merck
  • Comparative effects of fipronil and its metabolites sulfone and desulfinyl on the isolated rat liver mitochondria.
Comparative effects of fipronil and its metabolites sulfone and desulfinyl on the isolated rat liver mitochondria.
Environmental toxicology and pharmacology (2015-07-06)
Marco A Tavares, Ivo D F Palma, Hyllana C D Medeiros, Marieli Guelfi, Andréia T Santana, Fábio E Mingatto
ABSTRACT
Fipronil is an insecticide extensively used to control pests in crops and animals. There are relates of poisoning due to exposure of fipronil in mammals and the liver has been suggested as potential target. In this study, we evaluated the effects of fipronil and its metabolites sulfone and desulfinyl on the bioenergetics, reactive oxygen species (ROS) production and calcium efflux from mitochondria isolated from rat liver. Fipronil (5-25 μM) inhibited state-3 respiration in mitochondria energized with glutamate plus malate, substrates of complex I of the respiratory chain and decreased the mitochondrial membrane potential resulting in inhibition of ATP synthesis. Fipronil also caused uncoupling in succinate-energized mitochondria and calcium efflux. The metabolites sulfone and desulfinyl also acted as mitochondrial inhibitors and uncouplers and caused calcium efflux, but with different potencies, being the sulfone the more potent one. These effects of fipronil and its metabolites on mitochondrial bioenergetics and calcium homeostasis may be related to toxic effects of the insecticide in the liver.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) Bioluminescent Assay Kit, for ATP quantitation
Pricing and availability is not currently available.