Merck
  • Identification of p-hydroxybenzyl alcohol, tyrosol, phloretin and its derivate phloridzin as tyrosinase substrates.
Identification of p-hydroxybenzyl alcohol, tyrosol, phloretin and its derivate phloridzin as tyrosinase substrates.
Bioorganic & medicinal chemistry (2015-04-29)
Carmen Vanessa Ortiz-Ruiz, Jose Berna, Maria Del Mar Garcia-Molina, Jose Tudela, Virginia Tomas, Francisco Garcia-Canovas
ABSTRACT
In recent years, the hydroxyalkylphenols p-hydroxybenzyl alcohol and tyrosol, and the compound phloretin and its derivate phloridzin have been described as inhibitors of the enzyme tyrosinase. When the monophenolase and the diphenolase activities of tyrosinase on its physiological substrates l-dopa and/or l-tyrosine are measured in the presence of these compounds, the rate of action of the enzyme decreases. These findings led to the identification of these compounds as inhibitors. However, these molecules show an unusual behavior as inhibitors of the enzyme indeed, in this study, we demonstrate that they are not true inhibitors but alternative substrates of the enzyme.
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)