Merck
  • Ischemia-modified albumin and advanced oxidation protein products as potential biomarkers of protein oxidation in Alzheimer's disease.
Ischemia-modified albumin and advanced oxidation protein products as potential biomarkers of protein oxidation in Alzheimer's disease.
Geriatrics & gerontology international (2014-10-28)
Esma Altunoglu, Gulcan Guntas, Fusun Erdenen, Esen Akkaya, Ibrahim Topac, Hulya Irmak, Himmet Derici, Hakan Yavuzer, Remise Gelisgen, Hafize Uzun
ABSTRACT
The aim of the present study was to determine the systemic levels of oxidative stress markers, such as ischemia-modified albumin (IMA), advanced oxidation protein products (AOPP), ferric reducing antioxidant power (FRAP) and the prooxidant-antioxidant balance (PAB), to clarify protein redox homeostasis in patients with Alzheimer's disease, and to compare them with mentally healthy persons of the same age. A total of 38 patients with Alzheimer's disease (AD) and 34 sex- and age-matched mentally healthy control subjects were included in this study. The patients had significantly higher AOPP, IMA and PAB in the patient group than in the control group (P = 0.004, P = 0.001, P = 0.007, respectively). The FRAP was significantly lower in the patients with AD than in the control subjects (P = 0.002), and according to the receiver operating characteristic curves, the IMA and AOPP areas are below the 0.700 receiver operating characteristic curve line (area under the curve 0.817 and 0.730, respectively; 95% CI 0.709-0.898 and 0.612-0.828, respectively). Increased IMA, AOPP and PAB, and decreased FRAP are likely to be results of oxidative stress, a condition in which an imbalance occurs between the production and inactivation of reactive oxygen species in AD. The IMA could be used for the better evaluation of clinical status, as well as the independent characteristic symptoms of AD, for the purposes of routine clinical laboratory analysis.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications