Merck
  • Additivity of the Specific Effects of Additives on Protein Phase Behavior.
Additivity of the Specific Effects of Additives on Protein Phase Behavior.
The journal of physical chemistry. B (2015-11-07)
Florian Platten, Jan Hansen, Johanna Milius, Dana Wagner, Stefan U Egelhaaf
ABSTRACT
Protein phase behavior and protein-protein interactions can be tuned by additives. We experimentally determined the phase behavior of lysozyme solutions, namely, the cloud-point temperature (CPT), in the presence of two additives, sodium chloride (NaCl) and guanidine hydrochloride (GuHCl). Their concentrations are chosen to maintain the secondary structure, as verified by CD spectroscopy. Our data indicate that the salts affect the CPT through electrostatic screening and salt-specific contributions. At high salt concentrations, the CPT is a linear function of the additive concentration for the salts NaCl and GuHCl as well as for a nonionic additive, glycerol, and a solvent, dimethyl sulfoxide (DMSO). Their molar temperature increments, which rank their specific effects on the CPT (NaCl > 0 > DMSO > glycerol > GuHCl), are found to be essentially independent of the protein concentration. In particular, the specific effects of NaCl and GuHCl in mixtures are found to be additive, indicating the absence of synergies or suppressions between both salts. Thus, molar temperature increments represent a characteristic measure for the specific effects of additives on protein interactions, which is easily accessible in lab experiments and which will help to characterize the effects of additives on protein interactions and phase behavior.
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
SAFC
Guanidine hydrochloride
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O