Merck
  • TMPRSS3 modulates ovarian cancer cell proliferation, invasion and metastasis.
TMPRSS3 modulates ovarian cancer cell proliferation, invasion and metastasis.
Oncology reports (2015-11-05)
Dan Zhang, Shuang Qiu, Qi Wang, Jianhua Zheng
ABSTRACT
Overexpression of transmembrane protease, serine 3 (TMPRSS3) has been detected in ovarian cancer. However, the molecular mechanisms of TMPRSS3 in ovarian cancer remain unclear. In the present study, we found that TMPRSS3 was significantly expressed in ovarian cancer cells. Overexpression of TMPRSS3 promoted the proliferation, invasion and migration of A2780 cells. Conversely, knockdown of TMPRSS3 in HO8910 cells inhibited the proliferation, invasion and migration. Furthermore, TMPRSS3 affected the expression levels of E-cadherin, vimentin and Twist. In addition, TMPRSS3 induced activation of ERK1/2 in ovarian cancer cells, and the ERK1/2 pathway was required for the TMPRSS3-mediated proliferation, invasion and migration of ovarian cancer cells. Finally, knockdown of TMPRSS3 inhibited ovarian cancer HO8910 cell growth and metastasis in vivo. Collectively, the present study suggests that TMPRSS3 plays a crucial role in the development and progression of ovarian cancer. Therefore, TMPRSS3 represents a potential therapeutic target of ovarian cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human TMPRSS4
Sigma-Aldrich
MISSION® esiRNA, targeting human TMPRSS3
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tmprss4