Merck
  • Promoter hypermethylation of membrane type 3 matrix metalloproteinase is associated with cell migration in colorectal adenocarcinoma.
Promoter hypermethylation of membrane type 3 matrix metalloproteinase is associated with cell migration in colorectal adenocarcinoma.
Cancer genetics (2015-05-24)
Ji Wook Moon, Jong-Ho Choi, Soo Kyung Lee, Yong Woo Lee, Jung Ok Lee, Nami Kim, Hye Jeong Lee, Jung Seon Seo, Jin Kim, Hyeon Soo Kim, Gi Jin Kim, Sun-Hwa Park
ABSTRACT
The gene MT3-MMP (also known as MMP16) encodes the membrane type 3 matrix metalloproteinase, which is a member of the matrix metalloproteinase (MMP) gene family. Several MMPs are associated with migration in colorectal cancer (CRC). However, the methylation status of the MT3-MMP promoter in CRC has not been reported. The methylation status and expression levels of MT3-MMP were investigated in primary tumor tissues and adjacent normal tissues in 105 patients with CRC, one normal colon cell line (CCD18Co), and three CRC cell lines (SW480, DLD-1, and LoVo) by quantitative methylation-specific PCR and real-time PCR. MT3-MMP was hypermethylated in 82 of 105 CRC tissues (78%), 30 of 105 adjacent normal tissues (29%), and two of 11 normal colon tissues (18%). MT3-MMP mRNA was significantly reduced in CRC compared with that in adjacent normal tissues (P < 0.05). The methylation-mediated downregulation of MT3-MMP was restored by treatment with 5-aza-2'-deoxycytidine in two CRC cell lines, and MT3-MMP promoter activity was significantly reduced by methylation. The knockdown of MT3-MMP induced cell migration, but overexpressed MT3-MMP reduced cell migration in CRC cells. These results demonstrate that the MT3-MMP promoter is frequently hypermethylated in CRC and that downregulation of MT3-MMP may be important for cell migration in CRC.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP16
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mmp16
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Methanol, NMR reference standard