Merck
  • Effects of oestrogen deficiency on the alveolar bone of rats with experimental periodontitis.
Effects of oestrogen deficiency on the alveolar bone of rats with experimental periodontitis.
Molecular medicine reports (2015-06-03)
Xin-Chen Xu, Hui Chen, Xi Zhang, Zan-Jing Zhai, Xu-Qiang Liu, Xin-Yi Zheng, Jun Zhang, An Qin, Er-Yi Lu
ABSTRACT
Periodontitis is an inflammatory disease characterized by loss of connective tissue and alveolar bone, and osteoporosis is a common disease characterized by a systemic impairment of bone mass and microarchitecture. To date, the association between periodontitis and osteoporosis has remained to be fully elucidated. In the present study, an experimental rat model of periodontitis was used to explore the effects of oestrogen deficiency‑induced osteoporosis on the maxillary alveolar bone. Forty‑four female, six‑month‑old Sprague‑Dawley rats were randomly divided into four groups: Control, ligature, ovariectomized (OVX), and OVX + ligature. One month after ovariectomy, rats in the ligature and OVX + ligature groups received ligatures on their first and second maxillary molars for 1 month. Fluorescent labelling was performed prior to sacrificing the animals. At the end of the experiment, the maxillae and serum were collected and subjected to micro‑computed tomography analysis, confocal laser‑scanning microscopic observation, Van Gieson's fuchsin staining, tartrate‑resistant acid phosphatase staining and ELISA. Ligatures slightly reduced the alveolar bone mineral density (BMD) and bone formation rate, but significantly reduced alveolar crest height (ACH). Ovariectomy reduced the alveolar BMD, impaired the trabecular structure, reduced the bone formation rate and increased the serum levels of bone resorption markers. Animals in the OVX + ligature group exhibited a lower alveolar BMD, a poorer trabecular structure, a reduced ACH, a lower bone formation rate and higher serum levels of bone resorption markers compared with those in the control group. The results of the present study showed that ovariectomy enhanced alveolar bone loss and reduced the ACH of rats with experimental periodontitis. Thus, post‑menopausal osteoporosis may influence the progression of periodontitis.
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis