Merck
  • Percolating hierarchical defect structures drive phase transformation in Ce1-x Gd x O2-x/2: a total scattering study.
Percolating hierarchical defect structures drive phase transformation in Ce1-x Gd x O2-x/2: a total scattering study.
IUCrJ (2015-08-26)
Marco Scavini, Mauro Coduri, Mattia Allieta, Paolo Masala, Serena Cappelli, Cesare Oliva, Michela Brunelli, Francesco Orsini, Claudio Ferrero
ABSTRACT
A new hierarchical approach is presented for elucidating the structural disorder in Ce1-x Gd x O2-x/2 solid solutions on different scale lengths. The primary goal of this investigation is to shed light on the relations between the short-range and the average structure of these materials via an analysis of disorder on the mesocopic scale. Real-space (pair distribution function) and reciprocal-space (Rietveld refinement and microstructure probing) analysis of X-ray powder diffraction data and electron spin resonance (ESR) investigations were carried out following this approach. On the local scale, Gd- and Ce-rich droplets (i.e. small regions a few Ć„ngstrƶms wide) form, exhibiting either a distorted fluorite (CeO2) or a C-type (Gd2O3) structure in the whole compositional range. These droplets can then form C-type nanodomains which, for Gd concentrations x Gd ā‰¤ 0.25, are embedded in the fluorite matrix. At the site percolation threshold p C for a cubic lattice (x Gd = p C ā‰ƒ 0.311), C-type nanodomains percolate inside each crystallite and a structural phase transformation is observed. When this occurs, the peak-to-peak ESR line width Ī”H pp shows a step-like behaviour, which can be associated with the increase in Gd-Gd dipolar interactions. A general crystallographic rationale is presented to explain the fluorite-to-C-type phase transformation. The approach shown here could be adopted more generally in the analysis of disorder in other highly doped materials.
Product Description
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm Ɨ 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Driā„¢, ACS reagent, ā‰„99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%