Merck
  • Upper gastrointestinal Kaposi's sarcoma in HIV-infected patients: ten years of endoscopy observation at a single Brazilian center.
Upper gastrointestinal Kaposi's sarcoma in HIV-infected patients: ten years of endoscopy observation at a single Brazilian center.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2015-09-17)
Rosamar Eulira Fontes Rezende, Rafael Lima Kahwage, Tarciana Vieira da Costa, Alcyone Artioli Machado, Mariângela Ottoboni Brunaldi, Rafael Kemp, José Luiz Pimenta Módena
ABSTRACT
Kaposi's sarcoma (KS) is the most common neoplasm among HIV-infected individuals. The frequency of involvement of KS in the gastrointestinal (GI) tract and the associated epidemiological, immune, endoscopic, and histopathological features in HIV-infected patients, were evaluated in this study. A review of the medical and endoscopy reports of 1428 HIV-infected patients, who had undergone upper GI endoscopy at the Endoscopy Service, Clinical Hospital, Faculty of Medicine of Ribeirão Preto between January 1999 and June 2009, was performed. Clinical, epidemiological, immunological, endoscopic, and histological data were collected. Twenty-seven (1.9%) patients were diagnosed with GI KS. Patients were predominantly male (81.5%). Sexual activity was the main route of HIV transmission (81.5%). Cutaneous involvement was noted in 21 patients (78%). Fifteen patients (55%) received highly active antiretroviral therapy for a mean duration of 12.6 weeks (range 2-52 weeks) before endoscopy. GI lesions were mainly found in the stomach (55%). Analysis of the immunohistochemical methods HHV8 LNA-1, CD31, and CD34 for the diagnosis of gastric KS indicated high agreement (kappa=0.63, 95% confidence interval 0.32-0.94). There was no relationship between CD4 levels (p=0.34) or HIV viral load (p=0.99) and HHV8 LNA-1 positivity in gastric KS. GI KS is an infrequent finding in patients with HIV infection. Among those with GI KS, 80% had concomitant skin lesions. Immunohistochemical methods for CD31, CD34, and LNA-1 were important tools in the diagnostic assessment of lesions suggestive of KS in the GI tract. Further studies are required to confirm these data, and the need for routine endoscopic investigation of the GI tract in HIV-infected patients with cutaneous KS should be assessed.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)