Merck
  • Stimulation of the RIG-I/MAVS Pathway by Polyinosinic:Polycytidylic Acid Upregulates IFN-β in Airway Epithelial Cells with Minimal Costimulation of IL-8.
Stimulation of the RIG-I/MAVS Pathway by Polyinosinic:Polycytidylic Acid Upregulates IFN-β in Airway Epithelial Cells with Minimal Costimulation of IL-8.
Journal of immunology (Baltimore, Md. : 1950) (2015-08-19)
Nurlan Dauletbaev, Maria Cammisano, Kassey Herscovitch, Larry C Lands
ABSTRACT
Pharmacological stimulation of the antiviral cytokine IFN-β in the airways may help to counter deleterious virus-induced exacerbations in chronic inflammatory lung diseases (asthma, chronic obstructive pulmonary disease, or cystic fibrosis). Polyinosinic-polycytidylic acid [poly(I:C)] is a known inducer of IFN-β but also costimulates an inflammatory response. The latter response is undesirable given the pre-existing airway inflammation in these diseases. The objective of our study was to identify conditions for poly(I:C) to selectively upregulate IFN-β in airway epithelial cells without a concomitant inflammatory response. The inflammatory response was gauged by production of the chemokine IL-8. Using cell lines and primary airway epithelial cells (both submerged and well-differentiated), we observed that pure poly(I:C) stimulated IFN-β mainly through the TLR3/TRIF pathway and IL-8 through an unidentified pathway. The magnitude of the IL-8 response stimulated by pure poly(I:C) matched or even exceeded that of IFN-β. Furthermore, this IL-8 response could not be pharmacologically downregulated without affecting IFN-β. In contrast, we show that stimulation of the RIG-I/MAVS pathway, such as when poly(I:C) is delivered intracellularly in a complex with liposomes or via nucleofection, selectively stimulates IFN-β with low IL-8 costimulation. The magnitude of IFN-β stimulation by liposome-encapsulated poly(I:C) is markedly diminished in well-differentiated cells. In conclusion, it is feasible to augment IFN-β production in airway epithelial cells without excessive costimulation of IL-8 if the RIG-I/MAVS pathway is stimulated, such as via liposomal delivery of poly(I:C). Better cytoplasmic delivery vehicles are needed to efficiently stimulate this pathway in well-differentiated cells.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map3k14
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K14
Sigma-Aldrich
Budesonide, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)