Merck
  • Exosomes derived from human mesenchymal stem cells confer drug resistance in gastric cancer.
Exosomes derived from human mesenchymal stem cells confer drug resistance in gastric cancer.
Cell cycle (Georgetown, Tex.) (2015-06-20)
Runbi Ji, Bin Zhang, Xu Zhang, Jianguo Xue, Xiao Yuan, Yongmin Yan, Mei Wang, Wei Zhu, Hui Qian, Wenrong Xu
ABSTRACT
Mesenchymal stem cells (MSCs) play an important role in chemoresistance. Exosomes have been reported to modify cellular phenotype and function by mediating cell-cell communication. In this study, we aimed to investigate whether exosomes derived from MSCs (MSC-exosomes) are involved in mediating the resistance to chemotherapy in gastric cancer and to explore the underlying molecular mechanism. We found that MSC-exosomes significantly induced the resistance of gastric cancer cells to 5-fluorouracil both in vivo and ex vivo. MSC-exosomes antagonized 5-fluorouracil-induced apoptosis and enhanced the expression of multi-drug resistance associated proteins, including MDR, MRP and LRP. Mechanistically, MSC-exosomes triggered the activation of calcium/calmodulin-dependent protein kinases (CaM-Ks) and Raf/MEK/ERK kinase cascade in gastric cancer cells. Blocking the CaM-Ks/Raf/MEK/ERK pathway inhibited the promoting role of MSC-exosomes in chemoresistance. Collectively, MSC-exosomes could induce drug resistance in gastric cancer cells by activating CaM-Ks/Raf/MEK/ERK pathway. Our findings suggest that MSC-exosomes have profound effects on modifying gastric cancer cells in the development of drug resistance. Targeting the interaction between MSC-exosomes and cancer cells may help improve the efficacy of chemotherapy in gastric cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCB1
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TBC1D9