Merck
  • Annexin A1 Preferentially Predicts Poor Prognosis of Basal-Like Breast Cancer Patients by Activating mTOR-S6 Signaling.
Annexin A1 Preferentially Predicts Poor Prognosis of Basal-Like Breast Cancer Patients by Activating mTOR-S6 Signaling.
PloS one (2015-05-23)
Anjana Bhardwaj, Nivetha Ganesan, Kazunoshin Tachibana, Kimal Rajapakshe, Constance T Albarracin, Preethi H Gunaratne, Cristian Coarfa, Isabelle Bedrosian
ABSTRACT
Annexin A1 (ANXA1) is an anti-inflammatory protein reported to play a role in cell proliferation and apoptosis, and to be deregulated in breast cancer. The exact role of annexin A1 in the biology of breast cancer remains unclear. We hypothesized that the annexin A1 plays an oncogenic role in basal subtype of breast cancer by modulating key growth pathway(s). By mining the Cancer Genome Atlas (TCGA)-Breast Cancer dataset and manipulating annexin A1 levels in breast cancer cell lines, we studied the role of annexin A1 in breast cancer and underlying signaling pathways. Our in-silico analysis of TCGA-breast cancer dataset demonstrated that annexin A1 mRNA expression is higher in basal subtype compared to luminal and HER2 subtypes. Within the basal subtype, patients show significantly poorer overall survival associated with higher expression of annexin A1. In both TCGA patient samples and cell lines, annexin A1 levels were significantly higher in basal-like breast cancer than luminal and Her2/neu-positive breast cancer. Stable annexin A1 knockdown in TNBC cell lines suppressed the mTOR-S6 pathway likely through activation of AMPK but had no impact on the MAPK, c-Met, and EGFR pathways. In a cell migration assay, annexin A1-depleted TNBC cells showed delayed migration as compared to wild-type cells, which could be responsible for poor patient prognosis in basal like breast cancers that are known to express higher annexin A1. Our data suggest that annexin A1 is prognostic only in patients with basal like breast cancer. This appears to be in part due to the role of annexin A1 in activating mTOR-pS6 pathway.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
MISSION® esiRNA, targeting human ANXA1
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Anxa1