Merck
  • All-trans retinoic acid induces p53-depenent apoptosis in human hepatocytes by activating p14 expression via promoter hypomethylation.
All-trans retinoic acid induces p53-depenent apoptosis in human hepatocytes by activating p14 expression via promoter hypomethylation.
Cancer letters (2015-04-02)
Shin-Hee Heo, Juri Kwak, Kyung Lib Jang
ABSTRACT
All-trans retinoic acid (ATRA), the most biologically active metabolite of vitamin A, has been extensively studied for the prevention and treatment of cancer; however, the underlying mechanism of its anti-cancer potential is still unclear. Here we found that ATRA induces apoptosis in p53-positive HepG2 cells, but not in p53-negative Hep3B cells. For this effect, ATRA activated p14 expression via promoter hypomethylation, resulting in ubiquitin-dependent degradation of mouse double minute 2 (MDM2) and subsequent stabilization of p53. The potential of ATRA to stabilize p53 was almost completely abolished by knock-down of p14 in HepG2 cells and was not observed in p14-negative A549 cells. Upregulation of p14 also abolished the self-regulatory potential of p53 to repress p14 expression via DNA methylation and transcriptionally activate MDM2 expression. The accumulated p53 then activated several apoptosis-related molecules, including Bax, PUMA, caspase-9, Bid, caspase-8, caspase-3, and PARP. Ectopic expression of DNA methyltransferase 1 almost completely abolished the potential of ATRA to activate the p14-MDM2-p53 pathway and induce p53-dependent apoptosis. Therefore, we conclude that ATRA induces p14 promoter hypomethylation to trigger apoptosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human FABP1
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human S100A9
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Sigma-Aldrich
MISSION® esiRNA, targeting human SUB1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdk2ap2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Robld3
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sub1
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required