Merck
  • Characterization of novel perylene diimides containing aromatic amino acid side chains.
Characterization of novel perylene diimides containing aromatic amino acid side chains.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-08-25)
Mohammed J Farooqi, Mark A Penick, Jessica Burch, George R Negrete, Lorenzo Brancaleon
ABSTRACT
Perylene diimide derivatives have attracted initial interest as industrial dyes. Recently, much attention has been focused on their strong π-π stacks resulting from the large PDI aromatic core. These PDI stacks have distinct optical properties, and provide informative models that could mimic light-harvesting systems and initial charge transfer typical of photosynthetic systems. The absorption property of PDI derivatives may be tuned from visible to near-infrared region by peripheral substitution. We have studied a new class of PDI derivatives with aryl substituents derived from the side chains of aromatic aminoacids (Tyrosine, Tryptophan and Phenylalanine). We have investigated their absorption and the fluorescence properties in a set of organic solvents and established their different tendencies to aggregate in solution despite their solubility. Most aggregation appears to be unordered. One PDI analogue (the one formed from Tyr) in Methanol, however, appears to form J-type aggregates. Based on our results the compounds appear to be promising for future investigations regarding the interaction of these dyes with biomolecules.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)