Merck
  • The orphan receptor NOR1 participates in isoprenaline-induced cardiac hypertrophy by regulating PARP-1.
The orphan receptor NOR1 participates in isoprenaline-induced cardiac hypertrophy by regulating PARP-1.
British journal of pharmacology (2015-01-28)
Xiao-Jun Feng, Hui Gao, Si Gao, Zhuoming Li, Hong Li, Jing Lu, Jiao-Jiao Wang, Xiao-Yang Huang, Min Liu, Jian Zou, Jian-Tao Ye, Pei-Qing Liu
ABSTRACT
The orphan nuclear receptor NOR1 belongs to the NR4A subfamily of the nuclear hormone receptor superfamily, and is involved in glucose and fat metabolism. However, its potential contribution to cardiovascular diseases remains to be assessed. Here, the roles of NOR1 in cardiac hypertrophy induced by isoprenaline and the underlying molecular mechanisms were investigated. NOR1 was expressed in cardiomyocytes treated with isoprenaline. After NOR1 overexpression or knockdown in neonatal rat cardiomyocytes, cellular hypertrophy was monitored by measuring cell surface area and the mRNA of hypertrophic biomarkers. Interactions between NOR1 and PARP-1 were investigated by co-immunoprecipitation. NOR1 expression and PARP-1 activity were measured in rats with cardiac hypertrophy induced by isoprenaline. Treatment with isoprenaline significantly up-regulated NOR1 expression and PARP-1 activity both in vivo and in vitro. Specific gene silencing of NOR1 attenuated isoprenaline-induced cardiomyocyte hypertrophy, whereas NOR1 overexpression exacerbated cardiac hypertrophy. We identified a physical interaction between NOR1 and PARP-1, which was enhanced by NOR1 transfection and thereby led to PARP-1 activation. Overexpression of NOR1, but not C293Y, a NOR1 mutant lacking the PARP-1 binding activity, increased cellular surface area and the mRNA levels of atrial natriuretic factor and brain natriuretic polypeptide, effects blocked by the PARP-1 inhibitor 3-aminobenzamide or siRNA for PARP-1. This is the first evidence that NOR1 was involved in isoprenaline-induced cardiac hypertrophy. The pro-hypertrophic effect of NOR1 can be partly attributed to its regulation of PARP-1 enzymic activity.
Sigma-Aldrich
MISSION® esiRNA, targeting human LRRD1
Sigma-Aldrich
PARP1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
PARP-1 human, recombinant, expressed in E. coli