Merck
  • Effects of Four Formulations of Prostaglandin Analogs on Eye Surface Cells. A Comparative Study.
Effects of Four Formulations of Prostaglandin Analogs on Eye Surface Cells. A Comparative Study.
PloS one (2015-06-13)
Fernando Pérez-Roca, Esther Rodrigo-Morales, Ingrid Garzón, Ana-Celeste Oliveira, Miguel-Ángel Martín-Piedra, Víctor Carriel, Ana-Isabel Ortiz-Pérez, Indalecio Sánchez-Montesinos, Antonio Campos, Miguel Alaminos
ABSTRACT
We evaluated the cytotoxic effects of four prostaglandin analogs (PGAs) used to treat glaucoma. First we established primary cultures of conjunctival stromal cells from healthy donors. Then cell cultures were incubated with different concentrations (0, 0.1, 1, 5, 25, 50 and 100%) of commercial formulations of bimatoprost, tafluprost, travoprost and latanoprost for increasing periods (5 and 30 min, 1 h, 6 h and 24 h) and cell survival was assessed with three different methods: WST-1, MTT and calcein/AM-ethidium homodimer-1 assays. Our results showed that all PGAs were associated with a certain level of cell damage, which correlated significantly with the concentration of PGA used, and to a lesser extent with culture time. Tafluprost tended to be less toxic than bimatoprost, travoprost and latanoprost after all culture periods. The results for WST-1, MTT and calcein/AM-ethidium homodimer-1 correlated closely. When the average lethal dose 50 was calculated, we found that the most cytotoxic drug was latanoprost, whereas tafluprost was the most sparing of the ocular surface in vitro. These results indicate the need to design novel PGAs with high effectiveness but free from the cytotoxic effects that we found, or at least to obtain drugs that are functional at low dosages. The fact that the commercial formulation of tafluprost used in this work was preservative-free may support the current tendency to eliminate preservatives from eye drops for clinical use.
Sigma-Aldrich
Latanoprost, ≥98% (HPLC)
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
17-Phenyl-tri-norprostaglandin F-ethyl amide, ≥95%, solid
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fluprostenol isopropyl ester, ≥98%, ethanol solution
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.