Merck
  • Fate of microbial metabolites of dietary polyphenols in rats: is the brain their target destination?
Fate of microbial metabolites of dietary polyphenols in rats: is the brain their target destination?
ACS chemical neuroscience (2015-04-22)
Mattia Gasperotti, Sabina Passamonti, Federica Tramer, Domenico Masuero, Graziano Guella, Fulvio Mattivi, Urska Vrhovsek
ABSTRACT
Different polyphenol compounds are ingested when consuming a serving of fruits rich in polyphenols, spanning from one-phenol hydroxybenzoic acid to more complex polymeric compounds. Only a minor quantity of the polyphenols (5-10%) is absorbed. The remainder reaches the colon and is extensively metabolized by gut microbiota to low-molecular weight metabolites. Their subsequent tissue distribution is still undefined, although these microbial metabolites are currently believed to play a role in human health and disease states. To fill this knowledge gap, we performed a pharmacokinetics experiment in which a single bolus of 23 polyphenol microbial metabolites (total 2.7 μmol) was administered intravenously to rats to reliably reproduce a physiological postabsorption situation. Tissues and urine were collected shortly thereafter (15 s to 15 min) and were analyzed by UHPLC-MS/MS to quantitatively track these compounds. Remarkably, the brain was found to be a specific target organ for 10 of the 23 polyphenol metabolites injected, which significantly increased in the treated animals. In most cases, their appearance in the brain was biphasic, with an early wave at 2 min (4 compounds) and a second wave starting at 5 min; at 15 min, 9 compounds were still detectable. Most compounds were excreted into the urine. The concentrations in the brain of the treated animals were compared against those of the control group by Student's t test, with p-values < 0.1 considered to be statistically significant. These findings provide new perspectives for understanding the role of diet on brain chemistry. Our experimental approach has enabled us to obtain rich metabolomics information from a single experiment involving a limited number of animals.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, ≥96.0% (T)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)