Merck
  • Metal free columns for determination of deoxynucleotide monophosphate by liquid chromatography/mass spectrometry and application to oligonucleotide.
Metal free columns for determination of deoxynucleotide monophosphate by liquid chromatography/mass spectrometry and application to oligonucleotide.
Journal of chromatography. A (2015-07-05)
Sachie Shibayama, Hiroshi Sakamaki, Taichi Yamazaki, Akiko Takatsu
ABSTRACT
We have developed a highly sensitive method for the analysis of deoxynucleotide monophosphates (dNMPs), which involves the use of liquid chromatography/mass spectrometry (LC/MS) and a new metal-free column. The new column solves the problem that the phosphate group in dNMPs interacts with the metal portion of the device or column. After optimization of the analytical conditions, the limits of detection (LODs) of dNMPs were from 5.4ng/g to 6.3ng/g. Those values were 10 times lower than the LODs of previous methods. We applied the method to the determination of the base composition and the quantification of 20-mer oligonucleotide. Despite use of a very small sample amount of 14.5ng, we were able to determine the base composition, and the result was consistent with theoretical values. We were also able to quantify the mass fraction of oligonucleotide with 8.2% expanded uncertainty (k=2). By means of the developed method, we were able to analyze dNMPs with high sensitivity as well as determine the base composition and quantify the mass fraction of oligonucleotide despite use of a small amount of sample.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
2′-Deoxyadenosine 5′-monophosphate, Sigma Grade, 98-100%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M