Merck
  • Flavonoids from the aerial parts of Houttuynia cordata attenuate lung inflammation in mice.
Flavonoids from the aerial parts of Houttuynia cordata attenuate lung inflammation in mice.
Archives of pharmacal research (2015-03-07)
Ju Hee Lee, Jongmin Ahn, Jin Woong Kim, Sang Gook Lee, Hyun Pyo Kim
ABSTRACT
The aerial parts of Houttuynia cordata used for treating inflammation-related disorders contain flavonoids as major constituents. Since certain flavonoids possess anti-inflammatory activity, especially in the lung, the pharmacological activities of H. cordata and the flavonoid constituents were evaluated using in vitro and in vivo models of lung inflammation. The 70 % ethanol extract of the aerial parts of H. cordata inhibited the production of inflammatory biomarkers IL-6 and NO in lung epithelial cells (A549) and alveolar macrophages (MH-S), respectively. And the same plant material, administered orally (100 and 400 mg/kg), significantly inhibited lung inflammatory response in a mouse model of lipopolysaccharide (LPS)-induced acute lung injury. From the extract, major flavonoids including afzelin, hyperoside and quercitrin were successfully isolated and they also attenuated LPS-induced lung inflammation in mice by oral administration. In particular, quercitrin showed most potent activity at 100 mg/kg. These results demonstrate for the first time that H. cordata and three flavonoid constituents have a therapeutic potential for treating lung inflammatory disorders.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
2-Amino-5,6-dihydro-6-methyl-4H-1,3-thiazine, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC