Merck
  • Rapid and Easy Histological Evaluation of Alveolar Human Bone Quality at Dental Implant Sites Using a Nondecalcified Frozen Cryofilm Section Technique: A Technical Report.
Rapid and Easy Histological Evaluation of Alveolar Human Bone Quality at Dental Implant Sites Using a Nondecalcified Frozen Cryofilm Section Technique: A Technical Report.
Implant dentistry (2015-06-04)
Yuichi Ito, Hiroshi Fujita, Miwa Kanou, Yasuko Takahashi-Nakagawa, Yoichiro Nakajima, Akihiro Sunano, Yoshihiro Kimura, Takaaki Ueno
ABSTRACT
The evaluation of bone quality at the site of the alveolar bone for a dental implant is very important. This study presents an easy technique for direct evaluation of alveolar bone quality using nondecalcified cryofilm frozen sections on human alveolar bone core samples. Core samples harvested from alveolar bone were immediately frozen in cooled hexanen and slowly cut using a disposable tungsten carbide blade; the sliced sections were collected with adhesive cryofilms. Staining was performed using von toluidine blue and von Kossa for microscopic observations. All core samples clearly showed bone structure components of cortical bone, trabecular bone, bone marrow, blood vessels, and bone-related cells. These results suggest the efficacy of a nondecalcified cryofilm frozen section technique for histological observation of surgical implant sites.
Sigma-Aldrich
1-Hexene, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Hexene, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Hexene, 97%
Pricing and availability is not currently available.