Merck
  • An Azine-Linked Covalent Organic Framework: Synthesis, Characterization and Efficient Gas Storage.
An Azine-Linked Covalent Organic Framework: Synthesis, Characterization and Efficient Gas Storage.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Zhongping Li, Yongfeng Zhi, Xiao Feng, Xuesong Ding, Yongcun Zou, Xiaoming Liu, Ying Mu
ABSTRACT
A azine-linked covalent organic framework, COF-JLU2, was designed and synthesized by condensation of hydrazine hydrate and 1,3,5-triformylphloroglucinol under solvothermal conditions for the first time. The new covalent organic framework material combines permanent micropores, high crystallinity, good thermal and chemical stability, and abundant heteroatom activated sites in the skeleton. COF-JLU2 possesses a moderate BET surface area of over 410 m(2)  g(-1) with a pore volume of 0.56 cm(3)  g(-1) . Specifically, COF-JLU2 displays remarkable carbon dioxide uptake (up to 217 mg g(-1) ) and methane uptake (38 mg g(-1) ) at 273 K and 1 bar, as well as high CO2 /N2 (77) selectivity. Furthermore, we further highlight that it exhibits a higher hydrogen storage capacity (16 mg g(-1) ) than those of reported COFs at 77 K and 1 bar.
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%