Merck
  • Combination comet/micronucleus assay validation performed by BioReliance under the JaCVAM initiative.
Combination comet/micronucleus assay validation performed by BioReliance under the JaCVAM initiative.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
Kamala Pant, Ljubica Krsmanovic, Shannon Wilson Bruce, Tawney Kelley, Mirna Arevalo, Samuel Atta-Safoh, Fekadu Debelie, Michelle L Klug La Force, Sandra Springer, Jamie Sly, Madhav Paranjpe, Timothy Lawlor, Marilyn Aardema
ABSTRACT
In the international validation study of the in vivo rat alkaline comet assay (comet assay), the Japanese Center for the Validation of Alternative Methods (JaCVAM) provided three coded chemicals to BioReliance, 1,3-dichloropropene, ethionamide and busulfan, to be tested in a combined in vivo comet/micronucleus assay. Induction of DNA damage (comet) in liver, stomach and jejunum (1,3-dichloropropene only) cells, and induction of MNPCEs in bone marrow, were examined in male Sprague-Dawley (Hsd:SD) rats following oral administration of the test chemical for three consecutive days. A dose range finding (DRF) test was performed with each chemical to determine the maximum tolerated dose (MTD). Based on the results of the DRF test; 1,3-dichloropropene was tested at 50, 100 and 200 mg/kg/day; ethionamide was tested at 125, 250 and 500 mg/kg/day, and busulfan was tested at 10, 20 and 40 mg/kg/day. The results indicated that 1,3-dichloropropene induced DNA damage only in liver cells at all three test article doses, while no effects were observed in the stomach and jejunum cells. Additionally, it did not increase MNPCEs in the bone marrow. 1,3-Dichloropropene was concluded to be negative in the MN assay but positive in the comet assay. Ethionamide did not induce DNA damage in liver. However, in stomach, statistically significant decreases (although still within historical range) in % tail DNA at all test article doses compared to the vehicle control were observed. There was no increase in MNPCEs in the bone marrow. Thus, ethionamide was concluded to be negative in the comet/MN combined assay. Busulfan did not induce DNA damage in any of the organs tested (liver and stomach) but it did induce a significant increase in MNPCEs in the bone marrow. Busulfan was concluded to be negative in the comet assay but positive in the MN assay.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%