Merck
  • AMP-activated kinase (AMPK) regulates activity of HER2 and EGFR in breast cancer.
AMP-activated kinase (AMPK) regulates activity of HER2 and EGFR in breast cancer.
Oncotarget (2015-07-06)
Teraneh Z Jhaveri, Juhyung Woo, Xiaobin Shang, Ben Ho Park, Edward Gabrielson
ABSTRACT
AMP-activated Protein Kinase (AMPK) activity retards growth of many types of cancers. Investigating effects of AMPK activation on breast cancer cell signaling and survival, we found that breast cancer cell lines with amplification and over-expression of HER2 or EGFR are 2- to 5-fold more sensitive to cytotoxic effects of AICAR, a canonical pharmacological activator of AMPK, than breast cancer cell lines lacking HER2 or EGFR overexpression. Paralleling effects on cell survival, AICAR leads to dose- and time-dependent inhibition of HER2 and EGFR in HER2-amplified breast cancer cells, with activation of AMPK and suppression of HER2/ EGFR activity preceding commitment to cell death. Transfection of constitutively active AMPKα also leads to decreased HER2 and EGFR phosphorylation, reduced downstream signaling associated with these receptor tyrosine kinases (RTKs), and reduced breast cancer cell growth, confirming effects of AMPK activity on HER2/ EGFR. Ensuing co-immunoprecipitation experiments demonstrated an interaction of HER2 with AMPK and an in vitro phosphorylation assay found that HER2 and EGFR contain sequences that are potential substrates for AMPK. Our results lead us to postulate that AMPK regulates HER2 and EGFR activity in HER2-amplified breast cancer cells and thus activation of AMPK might provide therapeutic benefit in such cancers.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKAA2
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Prkaa2