Merck
  • Electrochemiluminescence DNA sensor array for multiplex detection of biowarfare agents.
Electrochemiluminescence DNA sensor array for multiplex detection of biowarfare agents.
Analytical and bioanalytical chemistry (2015-06-24)
Anna-Maria Spehar-Délèze, Rainer Gransee, Sergio Martinez-Montequin, Diego Bejarano-Nosas, Samuel Dulay, Sandra Julich, Herbert Tomaso, Ciara K O'Sullivan
ABSTRACT
Development of a fully automated electrochemiluminescence (ECL) DNA assay for multiplex detection of six biowarfare agents is described. Aminated-DNA capture probes were covalently immobilised on activated-carbon electrodes and subsequently hybridised to target strands. Detection was achieved via a sandwich-type assay after Ru(bpy)3(2+)-labelled reporter probes were hybridised to the formed probe-target complexes. The assay was performed in an automated microsystem in a custom designed ECL detection box with integrated fluidics, electronics,and movable photomultiplier detector. The obtained limits of detection were 0.6-1.2 nmol L(-1) for six targets ranging from 50 to 122 base pairs in size, with linear range 1-15 nmol L(-1). Non-specific adsorption and cross-reactivity were very low. Detection of six targets on a single chip was achieved with subnanomolar detection limits.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Tripropylamine, ≥98%
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading