Merck
  • Insertion, elimination and isomerisation of olefins at alkylaluminium hydride: an experimental and theoretical study.
Insertion, elimination and isomerisation of olefins at alkylaluminium hydride: an experimental and theoretical study.
Dalton transactions (Cambridge, England : 2003) (2015-06-05)
Nandita M Weliange, David S McGuinness, Michael G Gardiner, Jim Patel
ABSTRACT
The insertion, elimination and isomerisation of octenes with di-n-octylaluminium hydride [HAl(Oct)2], tri-n-octylaluminium [Al(Oct)3] and sec-octylaluminium species have been studied as individual steps in a putative aluminium based contrathermodynamic olefin isomerisation process. While elimination of 1-octene from [Al(Oct)3] is energetically unfavourable, the process is driven by high temperature vacuum distillation, leading to very high selectivity to 1-octene (>97%). At high conversions the [HAl(Oct)2] so obtained exists predominately as hydride-bridged cyclic oligomers, whereas at low conversion the mixed alkyl/hydride-bridged dimer [(Oct)2Al(μ-H)(μ-Oct)Al(Oct)2] is the major species. Di-n-octylaluminium hydride recovered after olefin elimination may be recycled and is active toward re-insertion of octenes. Internal octenes (cis- and trans-2-, 3- and 4-octene) only partially insert however, and even after prolonged heating there is no significant secondary to primary alkyl isomerisation evident.
Sigma-Aldrich
Triisobutylaluminum solution, 1.0 M in hexanes
Pricing and availability is not currently available.