Merck
  • Eryptosis Indices as a Novel Predictive Parameter for Biocompatibility of Fe3O4 Magnetic Nanoparticles on Erythrocytes.
Eryptosis Indices as a Novel Predictive Parameter for Biocompatibility of Fe3O4 Magnetic Nanoparticles on Erythrocytes.
Scientific reports (2015-11-06)
Qian Ran, Yang Xiang, Yao Liu, Lixin Xiang, Fengjie Li, Xiaojun Deng, Yanni Xiao, Li Chen, Lili Chen, Zhongjun Li
ABSTRACT
Fe3O4 magnetic nanoparticles (Fe3O4-MNPs) have been widely used in clinical diagnosis. Hemocompatibility of the nanoparticles is usually evaluated by hemolysis. However, hemolysis assessment does not measure the dysfunctional erythrocytes with pathological changes on the unbroken cellular membrane. The aim of this study is to evaluate the use of suicidal death of erythrocytes (i.e. eryptosis indices) as a novel predictive and prognostic parameter, and to determine the impact of Fe3O4-MNPs on cellular membrane structure and the rheology properties of blood in circulation. Our results showed that phosphatidylserine externalization assessment was significantly more sensitive than classical hemolysis testing in evaluating hemocompatibility. Although no remarkable changes of histopathology, hematology and serum biochemistry indices were observed in vivo, Fe3O4-MNPs significantly affected hemorheology indices including erythrocyte deformation index, erythrocyte rigidity index, red blood cell aggregation index, and erythrocyte electrophoresis time, which are related to the mechanical properties of the erythrocytes. Oxidative stress induced calcium influx played a critical role in the eryptotic activity of Fe3O4-MNPs. This study demonstrated that Fe3O4-MNPs cause eryptosis and changes in flow properties of blood, suggesting that phosphatidylserine externalization can serve as a predictive parameter for hemocompatibility assay.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SAFC
HEPES
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester, BioReagent, suitable for fluorescence, mixture of isomers, ≥80% (HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)