Merck
  • Identification of Arabidopsis thaliana NRT1/PTR FAMILY (NPF) proteins capable of transporting plant hormones.
Identification of Arabidopsis thaliana NRT1/PTR FAMILY (NPF) proteins capable of transporting plant hormones.
Journal of plant research (2015-03-25)
Yasutaka Chiba, Takafumi Shimizu, Shinya Miyakawa, Yuri Kanno, Tomokazu Koshiba, Yuji Kamiya, Mitsunori Seo
ABSTRACT
NRT1/PTR FAMILY (NPF) proteins were originally identified as nitrate or di/tri-peptide transporters. Recent studies revealed that this transporter family also transports the plant hormones auxin (indole-3-acetic acid), abscisic acid (ABA), and gibberellin (GA), as well as secondary metabolites (glucosinolates). We developed modified yeast two-hybrid systems with receptor complexes for GA and jasmonoyl-isoleucine (JA-Ile), to detect GA and JA-Ile transport activities of proteins expressed in the yeast cells. Using these GA and JA-Ile systems as well as the ABA system that we had introduced previously, we determined the capacities of Arabidopsis NPFs to transport these hormones. Several NPFs induced the formation of receptor complexes under relatively low hormone concentrations. Hormone transport activities were confirmed for some NPFs by direct analysis of hormone uptake of yeast cells by liquid chromatography-tandem mass spectrometry. Our results suggest that at least some NPFs could function as hormone transporters.
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Gibberellic acid, suitable for plant cell culture, BioReagent, ≥90% gibberellin A3 basis (of total gibberellins.)
Pricing and availability is not currently available.
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis
Pricing and availability is not currently available.
Sigma-Aldrich
Gibberellin, 80% gibberellin A3 basis (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Pricing and availability is not currently available.