Merck
  • Phenolic profile and antioxidant capacity of the principal apples produced in Brazil.
Phenolic profile and antioxidant capacity of the principal apples produced in Brazil.
International journal of food sciences and nutrition (2013-01-31)
Danianni Marinho Zardo, Karolline Marques Silva, Sylvain Guyot, Alessandro Nogueira
ABSTRACT
This study evaluated the phenols of the Gala, Fuji and Golden Delicious varieties, which make up 95% of Brazilian production. The phenolic profiles (whole fruit) were determined by high pressure liquid chromatography, total phenols were determined using the Folin-Ciocalteau method and antioxidant capacity by the FRAP method (whole fruit, skin, pulp and centre). The Golden Delicious had the highest phenol content (408 mg/fruit) compared to Fuji (194 mg/fruit) and Gala (162 mg/fruit), and the antioxidant capacity of the Golden Delicious was 2.5 and 3.6 times higher than that found in the Fuji and Gala, respectively. The phenolic profile for the three varieties showed 5-caffeoylquinic acid, ( - )-epicatechin, procyanidin B2 and phloridzin as major components with procyanidins as the predominant class and quercetin as the minority. The consumption of apple pulp may provide 48-78% of the antioxidant capacity; however, if the peel is ingested, it may increase to 79-89%. The type of apple variety and the parts ingested are factors that must be taken into consideration in consumption.
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Pricing and availability is not currently available.