Merck
  • Substrate specificity of human glutamine transaminase K as an aminotransferase and as a cysteine S-conjugate beta-lyase.
Substrate specificity of human glutamine transaminase K as an aminotransferase and as a cysteine S-conjugate beta-lyase.
Archives of biochemistry and biophysics (2008-03-18)
Arthur J L Cooper, John T Pinto, Boris F Krasnikov, Zoya V Niatsetskaya, Qian Han, Jianyong Li, David Vauzour, Jeremy P E Spencer
ABSTRACT
Rat kidney glutamine transaminase K (GTK) exhibits broad specificity both as an aminotransferase and as a cysteine S-conjugate beta-lyase. The beta-lyase reaction products are pyruvate, ammonium and a sulfhydryl-containing fragment. We show here that recombinant human GTK (rhGTK) also exhibits broad specificity both as an aminotransferase and as a cysteine S-conjugate beta-lyase. S-(1,1,2,2-Tetrafluoroethyl)-l-cysteine is an excellent aminotransferase and beta-lyase substrate of rhGTK. Moderate aminotransferase and beta-lyase activities occur with the chemopreventive agent Se-methyl-l-selenocysteine. l-3-(2-Naphthyl)alanine, l-3-(1-naphthyl)alanine, 5-S-l-cysteinyldopamine and 5-S-l-cysteinyl-l-DOPA are measurable aminotransferase substrates, indicating that the active site can accommodate large aromatic amino acids. The alpha-keto acids generated by transamination/l-amino acid oxidase activity of the two catechol cysteine S-conjugates are unstable. A slow rhGTK-catalyzed beta-elimination reaction, as measured by pyruvate formation, was demonstrated with 5-S-l-cysteinyldopamine, but not with 5-S-l-cysteinyl-l-DOPA. The importance of transamination, oxidation and beta-elimination reactions involving 5-S-l-cysteinyldopamine, 5-S-l-cysteinyl-l-DOPA and Se-methyl-l-selenocysteine in human tissues and their biological relevance are discussed.