Merck
  • A convenient synthesis of 2-imino-1,3-thiazine-4-one derivatives from the reaction of primary amines and phenyl isothiocyanate in the presence of acryloyl chloride.
A convenient synthesis of 2-imino-1,3-thiazine-4-one derivatives from the reaction of primary amines and phenyl isothiocyanate in the presence of acryloyl chloride.
Combinatorial chemistry & high throughput screening (2014-03-19)
Amin Zolali, Farough Nasiri, Sonia Omid-Niakan
ABSTRACT
An efficient, one-pot and three-component synthesis of biologically important heterocyclic compounds is described from the reaction of primary amines and phenyl isothiocyanate in the presence of acryloyl chloride at room temperature without the need to use any catalyst.
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Pricing and availability is not currently available.