Merck
  • 2-methyltetrahydrofuran--lithium hexafluoroarsenate: a superior electrolyte for the secondary lithium electrode.
2-methyltetrahydrofuran--lithium hexafluoroarsenate: a superior electrolyte for the secondary lithium electrode.
Science (New York, N.Y.) (1979-05-04)
V R Koch, J H Young
ABSTRACT
Rechargeable, high energy density lithium batteries require an efficient lithium electrode. Earlier work with electrolytes based on propylene carbonate, methyl acetate, and tetrahydrofuran yielded poor lithium electrode cycling efficiencies because of electrolyte reduction by lithium. Solutions of lithium hexafluoroarsenate in 2-methyltetrahydrofuran are found to be remarkably stable toward lithium, resulting in cycling efficiencies that approach 98 percent. The ability of 2-methyltetrahydrofuran to resist reduction by lithium is thought to be based on the position of its lowest unfilled molecular orbital relative to that of tetrahydrofuran.
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, anhydrous, ≥99%, Inhibitor-free
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, anhydrous, ≥99.0%, contains 250 ppm BHT as stabilizer
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, ReagentPlus®, ≥99.5%, contains 150-400 ppm BHT as stabilizer