Merck
  • Human brain proteins showing neuron-specific interactions with γ-secretase.
Human brain proteins showing neuron-specific interactions with γ-secretase.
The FEBS journal (2015-04-22)
Mitsuhiro Inoue, Ji-Yeun Hur, Takahiro Kihara, Yasuhiro Teranishi, Natsuko G Yamamoto, Taizo Ishikawa, Birgitta Wiehager, Bengt Winblad, Lars O Tjernberg, Sophia Schedin-Weiss
ABSTRACT
The transmembrane protease complex γ-secretase is a key enzyme in Alzheimer disease pathogenesis as it liberates the neurotoxic amyloid β-peptide (Aβ); however, the mechanism of regulation of its activity in various cell types and subcellular compartments is largely unknown. Several γ-secretase inhibitors have been developed, but none have been released due to side-effects that appear to arise from reduced processing of Notch, one of many γ-secretase substrates. Hence, it is desirable to specifically inhibit Aβ production. In our previous studies, we have identified several γ-secretase-associated proteins (GSAPs) from brain, which affect Aβ production without having any major effects on Notch processing. In the present study using detergent-resistant membranes prepared from brain, we have identified four GSAPs that affect Aβ production to a greater extent than Notch processing. We evaluated the interaction between GSAPs and γ-secretase in various cell types and their mRNA expression in various human organs. Using an in situ proximity ligation assay, we demonstrated that many GSAPs showed considerably greater interaction with γ-secretase in neurons than in human embryonic kidney cells stably over-expressing APP, and showed that several GSAPs are highly expressed in human brain. This study underscores the importance of studying protein-protein interactions in relevant cell types, and suggests that reducing Aβ production by interfering with brain- or neuron-specific γ-secretase/GSAP interactions may reduce the risk of unwanted side-effects associated with treatment of Alzheimer disease.
Sigma-Aldrich
Monoclonal Anti-NTRK2 antibody produced in mouse, clone 4D3-F10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Nicastrin Antibody, ascites fluid, clone 9C3, Chemicon®