Merck
Sugars including erythronic and threonic acids in human aqueous humour.
Current eye research (1999-07-27)
J J Harding, P C Hassett, K C Rixon, A J Bron, D J Harvey
ABSTRACT
Sugars in the aqueous humour of the eye serve both as a source of nutrients to the lens and other anterior ocular tissues, and potentially as an indicator of waste products from these tissues. In this work we intended to measure the levels of sugars in human blood and aqueous humour from cataract patients with and without diabetes. After initial results we decided to identify an unknown sugar component. Sugars were measured by hplc. The unknown sugar peak was identified by gas chromatography/mass spectrometry Very little fructose and sorbitol were found. Glucose levels were higher in both blood and aqueous from diabetic patients. During these analyses we found a major component that did not correspond to any sugar reported previously in aqueous humour. This was identified as a mixture of threonic and erythronic acids. Glucose levels increase in human aqueous humour in diabetes without markedly raised levels of sorbitol or fructose. Erythronic and threonic acids are normal components of aqueous humour and blood. They may be derived from glycated proteins or from degradation of ascorbic acid.
Sigma-Aldrich
D-Erythronic acid potassium salt, ≥97.0% (GC)