Merck
  • A glucose/hydrogen peroxide biofuel cell that uses oxidase and peroxidase as catalysts by composite bulk-modified bioelectrodes based on a solid binding matrix.
A glucose/hydrogen peroxide biofuel cell that uses oxidase and peroxidase as catalysts by composite bulk-modified bioelectrodes based on a solid binding matrix.
Bioelectrochemistry (Amsterdam, Netherlands) (2002-05-16)
A Pizzariello, M Stred'ansky, S Miertus
ABSTRACT
An improved composite bulk-modified bioelectrode setup based on a solid binding matrix (SBM) has been used to develop a glucose/hydrogen peroxide biofuel cell. Fuel is combined through a catalytically promoted reaction with oxygen into and oxidized species and electricity. The present work explores the feasibility of a sugar-feed biofuel cell based on SBM technology. The biofuel cell that utilizes mediators as electron transporters from the glucose oxidation pathway of the enzyme directly to electrodes is considered in this work. The anode was a glucose oxidase (GOx, EC 1.1.3.4)/ferrocene-modified SBM/graphite composite electrode. The cathode was a horseradish peroxidase (HRP, EC 1.11.1.7)/ferrocene-modified SBM/graphite composite electrode. The composite transducer material was layered on a wide polymeric surface to obtain the biomodified electrodic elements, anodes and cathodes and were assembled into a biofuel cell using glucose and H(2)O(2) as the fuel substrate and the oxidizer. The electrochemical properties and the characteristics of single composite bioelectrodes are described. The open-circuit voltage of the cell was 0.22 V, and the power output of the cell was 0.15 microW/cm(2) at 0.021 V. The biofuel cell proved to be stable for an extended period of continuous work (30 days). The reproducibility of the biotransducers fabrication was also investigated. In addition, an application of presented biofuel cell, e.g. the use of hydrolyzed corn syrup as renewable biofuels, was discussed.
Sigma-Aldrich
2-Hexadecanone, 98%
Pricing and availability is not currently available.